You searched for: Syringes for Chromatography
In liquid chromatography, the syringe functions primarily as a pipette or liquid-transfer device that loads a sample loop. The same largely holds true for gas chromatography. The syringe generally does not take an active role in injection, which occurs only after the sample has been displaced from the syringe. These syringes are designed for the highest performance in applications such as liquid or gas chromatography and the handling of corrosive gases and liquids, radioactive materials, and sterile solutions. Syringes are gas-tight and leak-proof.
GC Syringes for TriPlus™ RSH Autosampler, Thermo Scientific
Supplier: Thermo Scientific
Achieve accurate, precise, and reproducible injections easily using Thermo Scientific™ GC Syringes for Thermo Scientific™ TriPlus™ RSH Autosampler
Expand 1 Items
WHEATON® MicroLiter Dynamic Load and Wash (DLW) Syringe and Needles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Syringe for chromatography.
Expand 1 Items
GC Syringes for Thermo Scientific TriPlus RSH Autosampler, Thermo Scientific
Supplier: Thermo Scientific
Gas Chromatography Syringe 1uL Removable Needle, 57mm, 23 Gauge, Cone Tip, for use with Thermo Scientific Triplus RSH Autosampler
Expand 35 Items
Disposable Syringes, MicroLiter, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
MicroLiter's syringes feature polypropylene housings that are solvent-resistant and clearly marked for media and pore size.
Expand 9 Items
Manual GC Syringes, Thermo Scientific
Supplier: Thermo Scientific
GC Syringes for manual injections.
Expand 16 Items
KIMBLE® Plastic Luer Slip Micro Syringe, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
The Plastic Luer Slip Micro Syringes work for chromatography, nuclear medicine, HPLC sample preparation, and many laboratory procedures.
Expand 1 Items
GC Syringes for CTC Instruments, Thermo Scientific
Supplier: Thermo Scientific
GC Autosampler syringes for use with CTC GC instruments
Expand 11 Items
GC Syringes for Thermo Scientific Instruments, Thermo Scientific
Supplier: Thermo Scientific
GC Autosampler syringes for use with Thermo Scientific GC instruments
Expand 25 Items
Disposable Chromatography Syringe, Microsolv
Supplier: MicroSolv Technology Corp
These syringes are used in chromatography and dissolution testing
Expand 12 Items
HPLC Syringes, Thermo Fisher Scientific
Supplier: Thermo Scientific
These HPLC syringes are standard fixed needle syringes for Rheodyne/Valco™ injectors.
Expand 2 Items
Hamilton Syringes for CTC Analytics Sampler Handlers, MicroLiter
Supplier: DWK Life Sciences (KIMBLE)
Microliter Analytical Supplies Has A Full Line Of Lc And Gc Chromatography Consumables And Can Help You Determine The Best Products For Your Application.Keywords: Syringe 100Ul 22G Pt 3 X-Type
Expand 23 Items
WHEATON® MicroLiter Dynamic Load and Wash (DLW) Syringe Holder with Syringe, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Syringe holder with syringe for chromatography.
Expand 1 Items
GC Syringes for Agilent Instruments, Thermo Scientific
Supplier: Thermo Scientific
GC Autosampler syringes for use with Agilent GC instruments.
Expand 12 Items
Manual GC Syringes for GC Autosampler, Thermo Scientific
Supplier: Thermo Scientific
Achieve accurate, precise, and reproducible manual injections easily using Thermo Scientific™ Manual GC Syringes
Expand 2 Items
Syringes for Dionex HPLC Autosamplers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Agilent syringes for Dionex autosamplers ensure accurate and effective sampling.
Expand 2 Items
Hamilton Syringe Adapter Kits, MicroLiter
Supplier: DWK Life Sciences (KIMBLE)
The design of the C-line is based on the improvements employed in CTC's X-Type syringe
Expand 22 Items
Captiva Disposable Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Syringe, Captiva Disposable, 5 mL, Polypropylene
Expand 3 Items
CrossLab Gas Tight Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Gas tight syringes for dispensing gases and liquids.
Expand 1 Items
CTC Smart Autosampler Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Agilent now offers Smart Syringes compatible with your PAL3 Autosampler!
Expand 16 Items
CrossLab Color Coded Syringes for Manual Injection Valves, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Many Agilent manual syringes for GC have color-coded barrels to facilitate volume selection.
Expand 14 Items
GC Autosampler Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Agilent syringes are what you need for precise, effective, sample delivery from your GC autosampler.
Expand 107 Items
CrossLab Manual Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Agilent syringes for manual injection can be used with any type or brand of manual injection valve.
Expand 62 Items
AutoSystem Autosampler Syringe, Gas Tight, PerkinElmer
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Metal Plunger in Barrel with PTFE-Tipped Seal used for routine analysis.
Expand 1 Items
Microliter™ and Gastight® Agilent 7670, 7671 and 7672 ALS GC Syringes, Hamilton
Supplier: Hamilton
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Expand 2 Items
Hamilton Syringes for Shimadzu GC AOC-14/AOC-17/AOC-20, Hamilton
Supplier: Hamilton
These Microliter syringes incorporate a hand-fitted stainless-steel plunger with a finely bored glass syringe barrel, ensuring durability, and superior life. They are a high-quality and cost-effective replacement syringe for popular GC autosamplers, such as the Shimadzu AOC-14, AOC-17, and AOC-20.
Expand 1 Items
Microliter™ 900 Series Syringes, Hamilton
Supplier: Hamilton
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Expand 6 Items
Microliter™ and Gastight® Agilent 7693 GC Syringes, Hamilton
Supplier: Hamilton
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Expand 4 Items
Microliter™ PerkinElmer® Autosystem and Clarus 500 GC Syringes, Hamilton Company
Supplier: Hamilton
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Expand 2 Items
Microsyringes, PerkinElmer
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
These microsyringes are used for accurate and precise liquid delivery