SVL® Screw Thread Tubes, Chemglass
Supplier: Chemglass
Certificates
About this item
Screw thread tube for replacement of apparatus using the SVL® series of glass threads.
- Threads are tooled on one end of a 6" length of tubing
For use with solid caps with PTFE liner or open top cap and sealing ring for use as a tubing connector.
Customers Who Bought This Also Bought