ContactUsLinkComponent
Dickkopf-Related Protein 1, DKK1, Human ELISA, BioVendor
Dickkopf-Related Protein 1, DKK1, Human ELISA, BioVendor
Catalog # 101189-766
Supplier:  BioVendor
CAS Number:  
Dickkopf-Related Protein 1, DKK1, Human ELISA, BioVendor
Catalog # 101189-766
Supplier:  BioVendor
Supplier Number:  RD191207200R
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Primary antibody reactivity:
    Human
  • Description:
    Dickkopf-Related Protein 1 (DKK1) Human ELISA
  • Size:
    1 kit
  • Regulatory status:
    RUO
  • Cat. no.:
    101189-766
  • Supplier No.:
    CS091 VA
Specifications
About this item
Dickkopf-1 (Dkk-1) is encoded by the gene dickkopf together with other members of dickkopf protein family in vertebrates.
  • Calibration Range: 0.125–4 ng/mL
  • Limit of Detection: 0.032 ng/mL
  • Serum: 50uL/well
Dickkopf-1 (Dkk-1) is encoded by the gene dickkopf together with other members of dickkopf protein family in vertebrates, Dkk-2, –3, –4 and a distant family member soggy, sometimes also called Dickkopf-like protein 1 (DKKL1). Dickkopf name is derived from german dick=thick and kopf=head and this protein was independently found and characterised and named as „Sk“. Dkk-1/Sk is composed from 266 amino acids and has predicted molecular weight 25.8kDa. The protein has one N-glycosylation site at the N-terminus. Dkk proteins possess the signal sequence and have two typical cysteine rich domains. The second cysteine-rich domain is required for binding to Lrp6 and Kremen-2. Up to now known receptors are from the family Lrp (Lrp5/6) and Kremen 1 and 2. Dkks are modulating Wnt signalling. Their effect is mostly inhibitory with exception of the Dkk-2 which is activating Wnt-signalling. Wnt signalling pathways are shifting the cell proliferation, cell identity and cell polarity from embryonic to adult homeostasis. Wnt is forming the complex composed of seven-transmembrane receptor Frizzled (Fz) receptor and a lipoprotein-receptor related protein Lrp5 or Lrp6. This triple complex formation is stabilizing the beta-catenin and activating the pathway. Dkk-1 is inhibiting this process by its interaction with Lrp6 that is blocking the Wnt-Fz-Lrp complex formation. The Lrp6-Dkk-1 is entering the cell by endocytosis allowed by the coreceptor Kremen. Dkk-1 in mouse was found to be exprimed in bone, specifically in osteoblasts and osteocytes. The overexpression of the Dkk-1 in Xenopus is causing the ectopic head formation and blocation by anti-Dkk-1 antibodies lead to microcephaly. In mouse models, when missing the Dkk-1, there was found incomplete development of structures anterior to the midbrain and consequently this lead to the perinatal death. In limb development of Dkk-1 null mice there has been found syndactyly and polydactyly. Dkk-1 ELISA is estimating the Dkk-1 in serum or plasma thus can be used in the cancer research, especially bone and lung cancer, as well as in the Paget’s disease or in the problems with bone calcification.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.