Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Cidec (from E. coli)
Human Recombinant Cidec (from E. coli)
Catalog # 102830-686
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Cidec (from E. coli)
Catalog # 102830-686
Supplier:  BioVendor
Supplier Number:  RD172372100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    CIDEC|Fat-specific protein FSP27 homolog|Cell death activator CIDE-3|FSP27
  • Protein/peptide name:
    Cidec
  • Sequence:
    MKHHHHHHAS EYAMKSLSLL YPKSLSRHVS VRTSVVTQQL LSEPSPKAPR ARPCRVSTAD RSVRKGIMAY SLEDLLLKVR DTLMLADKPF FLVLEEDGTT VETEEYFQAL AGDTVFMVLQ KGQKWQPPSE QGTRHPLSLS HKPAKKIDVA RVTFDLYKLN PQDFIGCLNV KATFYDTYSL SYDLHCCGAK RIMKEAFRWA LFSMQATGHV LLGTSCYLQQ LLDATEEGQP PKGKASSLIP TCLKILQ
  • Cat. no.:
    102830-686
  • Supplier No.:
    RD172372100

Specifications

About this item

Cell death activator CIDE-3 regulates enlargement of lipid droplets. It is expressed mainly in adipocytes, intestine, heart, stomach, and weakly in brain, kidney and liver. Its overexpression in preadipocytes induces apoptosis.

Sequence: MKHHHHHHAS EYAMKSLSLL YPKSLSRHVS VRTSVVTQQL LSEPSPKAPR ARPCRVSTAD RSVRKGIMAY SLEDLLLKVR DTLMLADKPF FLVLEEDGTT VETEEYFQAL AGDTVFMVLQ KGQKWQPPSE QGTRHPLSLS HKPAKKIDVA RVTFDLYKLN PQDFIGCLNV KATFYDTYSL SYDLHCCGAK RIMKEAFRWA LFSMQATGHV LLGTSCYLQQ LLDATEEGQP PKGKASSLIP TCLKILQ

Other names: Cell death activator CIDE-3, Fat-specific protein FSP27 homolog, CIDEC, FSP27

Ordering information: For research use only.