Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Serglycin (from E. coli)
Human Recombinant Serglycin (from E. coli)
Catalog # 102830-674
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Serglycin (from E. coli)
Catalog # 102830-674
Supplier:  BioVendor
Supplier Number:  RD172368100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    P.PG|Secretory granule proteoglycan core protein|Hematopoietic proteoglycan core protein|SRGN|Platelet proteoglycan core protein
  • Protein/peptide name:
    Serglycin
  • Sequence:
    MKHHHHHHAS YPTRRARYQW VRCNPDSNSA NCLEEKGPMF ELLPGESNKI PRLRTDLFPK TRIQDLNRIF PLSEDYSGSG FGSGSGSGSG SGSGFLTEME QDYQLVDESD AFHDNLRSLD RNLPSDSQDL GQHGLEEDFM L
  • Cat. no.:
    102830-674
  • Supplier No.:
    RD172368100

Specifications

About this item

Serglycin was found to be among the most abundantly expressed proteins in adipocytes of epicardial adipose tissue in patients with coronary artery disease (CAD). It was demonstrated that TNFα induces expression and secretion of serglycin in adipocytes. Circulating serglycin might also participate in the mechanism of systemic vascular insult and atheromatous change. Serglycin is a dominant intracellular proteoglycan expressed by immune cells, where it interacts with numerous inflammatory mediators, such as proteases, chemokines, cytokines, and growth factors. Serglycin is implicated in their storage into the granules and their protection since they are secreted as complexes and delivered to their targets after secretion.

Sequence: MKHHHHHHAS YPTRRARYQW VRCNPDSNSA NCLEEKGPMF ELLPGESNKI PRLRTDLFPK TRIQDLNRIF PLSEDYSGSG FGSGSGSGSG SGSGFLTEME QDYQLVDESD AFHDNLRSLD RNLPSDSQDL GQHGLEEDFM L

Other names: Hematopoietic proteoglycan core protein, Platelet proteoglycan core protein, P.PG, Secretory granule proteoglycan core protein,, SRGN

Ordering information: For research use only.