Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Cyclophilin F (from E. coli)
Human Recombinant Cyclophilin F (from E. coli)
Catalog # 102830-668
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Cyclophilin F (from E. coli)
Catalog # 102830-668
Supplier:  BioVendor
Supplier Number:  RD172364100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    Peptidyl-prolyl cis-trans isomerase F mitochondrial|CyP-M|CypD|Mitochondrial cyclophilin|Rotamase F|PPIase F|CyP-D
  • Protein/peptide name:
    Cyclophilin F
  • Sequence:
    MKHHHHHHAS CSKGSGDPSS SSSSGNPLVY LDVDANGKPL GRVVLELKAD VVPKTAENFR ALCTGEKGFG YKGSTFHRVI PSFMCQAGDF TNHNGTGGKS IYGSRFPDEN FTLKHVGPGV LSMANAGPNT NGSQFFICTI KTDWLDGKHV VFGHVKEGMD VVKKIESFGS KSGRTSKKIV ITDCGQLS
  • Cat. no.:
    102830-668
  • Supplier No.:
    RD172364100

Specifications

About this item

Cyclophilins facilitate the process of protein folding. They catalyzes the cis-trans isomerization of proline imidic peptide bonds. Cyclophilin F is located in mitochondrion matrix. Cyclophilin F is involved in regulation of the mitochondrial permeability transition pore. It may activate or inhibit apoptosis or necrosis. It binds to an immunosuppressant cyclosporin.

Sequence: MKHHHHHHAS CSKGSGDPSS SSSSGNPLVY LDVDANGKPL GRVVLELKAD VVPKTAENFR ALCTGEKGFG YKGSTFHRVI PSFMCQAGDF TNHNGTGGKS IYGSRFPDEN FTLKHVGPGV LSMANAGPNT NGSQFFICTI KTDWLDGKHV VFGHVKEGMD VVKKIESFGS KSGRTSKKIV ITDCGQLS

Other names: Peptidyl-prolyl cis-trans isomerase F mitochondrial, PPIase F, CyP-D, CypD, Mitochondrial cyclophilin, CyP-M, Rotamase F

Ordering information: For research use only.