Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Cyclophilin B (from E. coli)
Human Recombinant Cyclophilin B (from E. coli)
Catalog # 102830-662
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Cyclophilin B (from E. coli)
Catalog # 102830-662
Supplier:  BioVendor
Supplier Number:  RD172360100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    S-cyclophilin|PPIB|PPIase B|SCYLP|CYPB|Peptidyl-prolyl cis-trans isomerase B|Rotamase B|CYP-S1
  • Protein/peptide name:
    Cyclophilin B
  • Sequence:
    MKHHHHHHAS DEKKKGPKVT VKVYFDLRIG DEDVGRVIFG LFGKTVPKTV DNFVALATGE KGFGYKNSKF HRVIKDFMIQ GGDFTRGDGT GGKSIYGERF PDENFKLKHY GPGWVSMANA GKDTNGSQFF ITTVKTAWLD GKHVVFGKVL EGMEVVRKVE STKTDSRDKP LKDVIIADCG KIEVEKPFAI AKE
  • Cat. no.:
    102830-662
  • Supplier No.:
    RD172360100

Specifications

About this item

Cyclophilins facilitate the process of protein folding. They catalyzes the cis-trans isomerization of proline imidic peptide bonds. Cyclophilin B is located in endoplasmic reticulum lumen. It is inhibited by an immunosuppressant cyclosporin.

Sequence: MKHHHHHHAS DEKKKGPKVT VKVYFDLRIG DEDVGRVIFG LFGKTVPKTV DNFVALATGE KGFGYKNSKF HRVIKDFMIQ GGDFTRGDGT GGKSIYGERF PDENFKLKHY GPGWVSMANA GKDTNGSQFF ITTVKTAWLD GKHVVFGKVL EGMEVVRKVE STKTDSRDKP LKDVIIADCG KIEVEKPFAI AKE

Other names: Peptidyl-prolyl cis-trans isomerase B, PPIase B, CYP-S1, Rotamase B, S-cyclophilin, SCYLP, PPIB, CYPB

Ordering information: For research use only.