Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Neuritin (from E. coli)
Human Recombinant Neuritin (from E. coli)
Catalog # 102830-656
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Neuritin (from E. coli)
Catalog # 102830-656
Supplier:  BioVendor
Supplier Number:  RD172358100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    NRN|NRN1
  • Protein/peptide name:
    Neuritin
  • Sequence:
    MKHHHHHHAS AGKCDAVFKG FSDCLLKLGD SMANYPQGLD DKTNIKTVCT YWEDFHSCTV TALTDCQEGA KDMWDKLRKE SKNLNIQGSL FELCGSGNG
  • Cat. no.:
    102830-656
  • Supplier No.:
    RD172358100

Specifications

About this item

Neuritin was identified as an important neurotrophin expressed in the developing nervous system. This protein plays multiple roles in the process of neural development, synaptic plasticity, and synaptic maturation.

Sequence: MKHHHHHHAS AGKCDAVFKG FSDCLLKLGD SMANYPQGLD DKTNIKTVCT YWEDFHSCTV TALTDCQEGA KDMWDKLRKE SKNLNIQGSL FELCGSGNG

Other names: NRN1, NRN

Ordering information: For research use only.