Order Entry
Canada
ContactUsLinkComponent
Human Recombinant ANGPTL6 (from E. coli)
Human Recombinant ANGPTL6 (from E. coli)
Catalog # 102830-638
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant ANGPTL6 (from E. coli)
Catalog # 102830-638
Supplier:  BioVendor
Supplier Number:  RD172306100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    AGF|Angiopoietin-like Protein 6|Angiopoietin-related Growth Factor
  • Protein/peptide name:
    ANGPTL6
  • Cat. no.:
    102830-638
  • Supplier No.:
    RD172306100

Specifications

About this item

Seven angiopoietin-like proteins (ANGPTLs) share the char-acteristic protein structure of the angiopoietin family (ANG), but differ in their inability to bind antiopoietin receptor, Tie-2. ANGPTL6 was originally named angiopoietin-related growth factor (AGF) having an N-terminal coiled-coil-like domain and a C-terminal fibrinogen-like domain, both of which are conserved in ANG. It is a circulating protein secreted by liver that induces angiogenesis by direct effect of epidermal ANGPTL6 on endothelial cells and proliferation of skin cells, and thereby promotes wound healing.

Sequence: MKHHHHHHAS RAGAPRCTYT FVLPPQKFTG AVCWSGPAST RATPEAANAS ELAALRMRVG RHEELLRELQ RLAAADGAVA GEVRALRKES RGLSARLGQL RAQLQHEAGP GAGPGADLGA EPAAALALLG ERVLNASAEA QRAAARFHQL DVKFRELAQL VTQQSSLIAR LERLCPGGAG GQQQVLPPPP LVPVVPVRLV GSTSDTSRML DPAPEPQRDQ TQRQQEPMAS PMPAGHPAVP TKPVGPWQDC AEARQAGHEQ SGVYELRVGR HVVSVWCEQQ LEGGGWTVIQ RRQDGSVNFF TTWQHYKAGF GRPDGEYWLG LEPVYQLTSR GDHELLVLLE DWGGRGARAH YDGFSLEPES DHYRLRLGQY HGDAGDSLSW HNDKPFSTVD RDRDSYSGNC ALYQRGGWWY HACAHSNLNG VWHHGGHYRS RYQDGVYWAE FRGGAYSLRK AAMLIRPLKL

Other names: ANGPTL6

Ordering information: For research use only.