Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant CRISP3 (from E. coli)
Catalog # 102830-632
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant CRISP3 (from E. coli)
Catalog # 102830-632
Supplier:  BioVendor
Supplier Number:  RD172262100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    Aeg2|Specific Granule Protein|Rd172262100-10Cysteine-Rich Secretory Protein|28 KDa|SGP28|CRS3
  • Protein/peptide name:
    CRISP3
  • Sequence:
    MKHHHHHHAS NEDKDPAFTA LLTTQTQVQR EIVNKHNELR RAVSPPARNM LKMEWNKEANAQKWANQC NYRHSNPKDR MTSLKCGENL YMSSASSSWS QAIQSWFDEY NDFDFGVGPK TPNAVVGHYT QVVWYSSYLV GCGNAYCPNQ KVLKYYYVCQ YCPAGNWANR LYVPYEQGAP CASCPDNCDD GLCTNGCKYE DLYSNCKSLK LTLTCKHQLV RDSCKASCNC SNSIY
  • Cat. no.:
    102830-632
  • Supplier No.:
    RD172262100

Specifications

About this item

Cysteine-rich secretory protein 3 (CRISP-3, SGP 28) belongs to the CRISP family of proteins having a PR-1 (pathogenesis related protein 1) domain at its N-terminal and ion channel regulatory (ICR) domain at its C-terminal connected by a hinge region. CRISP-3 transcripts are shown to be present predominantly in the prostate. Its expression is low in benign prostatic epithelium but is highly upregulated in the majority of high-grade prostatic intraepithelial neoplasia lesions and in most primary prostate tumours and metastases. Two independent studies have shown that CRISP-3 is one of the genes that are most often upregulated in prostate cancer. Human Prostate Secretory Protein (PSP94) has been shown to bind human CRISP-3 with very high affinity. Functional significance of this complex is not yet known. Yet, it was suggested, that involvement of the hinge region of CRISPs in interaction with PSP94 may affect the domain movement of CRISPs essential for the ion-channel regulatory activity resulting in inhibition of this activity.

Sequence: MKHHHHHHAS NEDKDPAFTA LLTTQTQVQR EIVNKHNELR RAVSPPARNM LKMEWNKEAA ANAQKWANQC NYRHSNPKDR MTSLKCGENL YMSSASSSWS QAIQSWFDEY NDFDFGVGPK TPNAVVGHYT QVVWYSSYLV GCGNAYCPNQ KVLKYYYVCQ YCPAGNWANR LYVPYEQGAP CASCPDNCDD GLCTNGCKYE DLYSNCKSLK LTLTCKHQLV RDSCKASCNC SNSIY

Other names: CRISP-3, Specific Granule Protein (28 KDa), CRS3, SGP28, Aeg2

Ordering information: For research use only.