Order Entry
Canada
ContactUsLinkComponent
Human Recombinant CRISP2 (from E. coli)
Human Recombinant CRISP2 (from E. coli)
Catalog # 102830-626
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant CRISP2 (from E. coli)
Catalog # 102830-626
Supplier:  BioVendor
Supplier Number:  RD172261100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    GAPDL5|TPX1|Cancer/testis antigen 36|CT36|Rd172261100-10Cysteine-Rich Secretory Protein|CRISP2 |Testis-specific protein TPX-1
  • Protein/peptide name:
    CRISP2
  • Molecular weight:
    26.32 kDa
  • Sequence:
    MKHHHHHHAS KDPAFTALLT TQLQVQREIV NKHNELRKAV SPPASNMLKM EWSREVTTNA QRWANKCTLQ HSDPEDRKTS TRCGENLYMS SDPTSWSSAI QSWYDEILDF VYGVGPKSPN AVVGHYTQLV WYSTYQVGCG IAYCPNQDSL KYYYVCQYCP AGNNMNRKNT PYQQGTPCAG CPDDCDKGLC TNSCQYQDLL SNCDSLKNTA GCEHELLKEK CKATCLCENK IY
  • Cat. no.:
    102830-626
  • Supplier No.:
    RD172261100

Specifications

About this item

Recombinant protein contains amino acids no. 22–243 of the protein acc.no. P16562 in UniProtKB. Total 232 AA. MW: 26.32 kDa (calculated). N-Terminal His-tag (10 extra AA)

Sequence: MKHHHHHHAS KDPAFTALLT TQLQVQREIV NKHNELRKAV SPPASNMLKM EWSREVTTNA QRWANKCTLQ HSDPEDRKTS TRCGENLYMS SDPTSWSSAI QSWYDEILDF VYGVGPKSPN AVVGHYTQLV WYSTYQVGCG IAYCPNQDSL KYYYVCQYCP AGNNMNRKNT PYQQGTPCAG CPDDCDKGLC TNSCQYQDLL SNCDSLKNTA GCEHELLKEK CKATCLCENK IY

Other names: CRISP-2, Cancer/testis antigen 36, CT36, Testis-specific protein TPX-1, CRISP2 ,GAPDL5, TPX1

Ordering information: For research use only.