Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Protein synonyms:HMGB1|High Mobility Group Protein|High mobility group protein 1
- Protein/peptide name:HMGB1
- Molecular weight:21.8 kDa
- Sequence:MKHHHHHHAS GKGDPKKPRG KMSSYAFFVQ TCREEHKKKH PDASVNFSEF SKKCSERWKT MSAKEKGKFE DMAKADKARY EREMKTYIPP KGETKKKFKD PNAPKRPPSA FFLFCSEYRP KIKGEHPGLS IGDVAKKLGE MWNNTAADDK QPYEKKAAKL KEKYEKDIYRAKGKPDKKGVVKAEKS
- Cat. no.:102830-620
- Supplier No.:RD172232100
Total 190 AA. MW: 21.8 kDa (calculated). UniProtKB acc.no. P09429 (Gly2-Ser181). N-terminal histag (10 extra AA). Protein identity confirmed by LC-MS/MS.
Sequence: MKHHHHHHAS GKGDPKKPRG KMSSYAFFVQ TCREEHKKKH PDASVNFSEF SKKCSERWKT MSAKEKGKFE DMAKADKARY EREMKTYIPP KGETKKKFKD PNAPKRPPSA FFLFCSEYRP KIKGEHPGLS IGDVAKKLGE MWNNTAADDK QPYEKKAAKL KEKYEKDIAA YRAKGKPDAA KKGVVKAEKS
Other names: HMGB1, High mobility group protein 1