Order Entry
Canada
Orders LinkContactUsLinkComponent
Rigid-Lock QUICKBERM® Lite, Justrite®

Rigid-Lock QUICKBERM® Lite, Justrite®

EnGuard™I, modified PVC coated fabric, 33L x 20.3W x 17.8H cm (13 x 8 x 7"), 243.8W x 121.9D x 20.3H cm (96 x 48 x 8"), 243.8L x 121.9W x 20.3H cm (96 x 48 x 8"), 602 L (159 gal.)
Catalog # 10861-160
Supplier:   Justrite
Supplier #:  28564
Rigid-Lock QUICKBERM® Lite, Justrite®
Catalog # 10861-160
Supplier:  Justrite
Rigid-Lock QUICKBERM® Lite, Justrite®
Rigid-Lock QUICKBERM® Lite, Justrite®

EnGuard™I, modified PVC coated fabric, 33L x 20.3W x 17.8H cm (13 x 8 x 7"), 243.8W x 121.9D x 20.3H cm (96 x 48 x 8"), 243.8L x 121.9W x 20.3H cm (96 x 48 x 8"), 602 L (159 gal.)

Price
930,32 $
New Customer Promotional Price
Log in for availability
Each (159US Gal)
Product Details & Documents

Portable, versatile containment berm deploys quickly for emergency spill response.

  • Leakproof and chemical resistance
  • Reinforced inside supports reduce sagging and prevent tripping hazards that can be caused by outside supports

Lightweight berm folds down easily to a small size for storage in trucks, emergency vehicles, and spill kits. Add optional grates to help reduce contact with contaminated liquids by personnel or equipment. Helps comply with EPA for containment and spill prevention (SPCC).

Specifications
  • Material:
    EnGuard™I, modified PVC coated fabric
  • Interior dimensions:
    243.8W x 121.9D x 20.3H cm (96 x 48 x 8")
  • Ext. dimensions:
    243.8L x 121.9W x 20.3H cm (96 x 48 x 8")
  • Sump capacity:
    602 L (159 gal.)
  • Folded size:
    33L x 20.3W x 17.8H cm (13 x 8 x 7")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.