Finnpipette™ BioControl Multi Channel Tip Cone Module, Thermo Scientific
Catalog # 46620-274
Supplier: Thermo Fisher Scientific
Specifications
- Volume:5 - 50 µl
- Description:16-Channel Tip Cone Module
- Cat. no.:46620-274
- Increments:0.5 µl
- Accuracy:±5.0 - 2.4%
- Supplier No.:2207030
Specifications
About this item
These are interchangeable with single channel biocontrol modules.
- Variable volume
- Autoclavable
- Easy to remove