About this item
Nucleic acid isolation, Mini-prep kit used to isolate bacterial, fungal, plant and animal genomic DNA from soil and other environmental samples.
- Pack Size: 50 preparations
- Harndess: Ceramic Spheres, Vickers 800; Silica Spheres, Knoop 515; Glass Bead, Knoop 515
- Specific Gravity: Ceramic Spheres, 4; Silica Spheres, 2.5; Glass Bead, 2.5
The FastDNA SPIN kt for Soil is designed to isolate bacterial, fungal, plant and animal genomic DNA from soil and other environmental samples. Soil is processed with a FastPrep® instrument and lysing matrix E tubes designed to efficiently lyse all microorganisms.
The released DNA is purified by a silica-based spin filter method and is suitable for PCR analysis and other downstream applications. Complete lysis of all organisms, including historically difficult sources such as eubacterial spores and endospores, gram positive bacteria, yeast, algae, nematodes and fungi is achieved within seconds using a FastPrep instrument and Lysing Matrix E tubes.
Cited in over 6000 scientific publications, the FastDNA SPIN kit for Soil delivers the highest DNA yield from any environmental sample including soil, sediment, sludge, compost, manure, rhizosphere or waste water. The FastDNA SPIN Kit for Soil contains lysing matrix tubes and all the reagents needed to extract DNA in a process that efficiently removes humic acids and other PCR inhibitors.
Kit contents: 50×2.0 ml tubes of lysing matrix E, 60 ml sodium phosphate buffer, 8 ml MT buffer, 25 ml of PPS solution, 66 ml binding matrix, 50 spin modules, 50 catch tubes, 12 ml concentrated SEWS-M, 20 ml DES, 200 µL BBS gel loading dye.
Reagent: SEWS-M - pH range: 6.4 to 6.8.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Prep size:Mini
- Refine target molecule:DNA preps
- Sample type:Plant samplesStoolBacteriaYeast/FungiWastewaterSoil
- Target molecule:DNA
- Storage conditions:15…30° C
- Instrument Compatibility:FastPrep-24™ or FastPrep-96™
- Sample size:Up to 500 mg per preparation