About this item
The FastPrep-24 5G instrument is a high-speed benchtop homogenizer offering the ultimate in speed and performance for the lysis of biological samples.
- Simultaneous homogenization of up to 24 samples takes place in 40 seconds or less
- Unique, optimized motion to disrupt cells
- Developed for difficult and resistant samples
Samples and buffers are added to a 2.0 ml lysing matrix tube containing specialized lysing matrix beads. The ergonomic design of the instrument ensures an easy loading of the sample tubes that remains securely sealed during the processing. The homogenization speed and duration times are manually programmed using a touch screen visual display. After setting the speed and time with the touch of a button, just push 'run', and in less than a minute the samples are completely lysed. The touch screen software based interface also includes a wide range of recommended programs designed to minimize optimization times.
The FastPrep-24 5G is the only available homogenizer with nine optional interchangeable adapters which allows a unique versatility in sample number, size and temperature conditions. All adapters stand stable on the benchtop and are commonly used as tube racks for sample storage at -20 °C or -80 °C. Frozen lysing matrix tubes loaded in the adapters are ready to be immediately processed in the FastPrep-24 5G with minimal hands-on manipulation. A wide range of specialized lysing matrix tubes containing beads of different materials, sizes and shapes have been tailored to guarantee thorough homogenization of any sample. High performance purification kits, when used in conjunction with the FastPrep-24 5G, provide simple, ready-to-use methods for the release and subsequent purification of DNA, RNA and proteins.
Suitable for all sample types; lysing highly contagious samples; lysis of yeast and fungi; used for isolation of PCR-ready genomic DNA from fecal samples; used for isolation of PCR-ready genomic DNA from fungi and bacteria samples; used for isolation of PCR-ready genomic DNA from soil samples; DNA & RNA purification; lysis of bacteria; lysis of soft tissues; isolation of genomic DNA; purification of DNA; cell lysis; used to lyse cell wall of yeast; break hard/brittle samples; used to isolate DNA from virtually any source.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76046-056
- Weight:
- 23.6 kg (52 lbs.)
- Tube compatibility:
- 2 ml tube; 4.5 ml tube; 15 ml tube; 50 ml tube
- Sample throughput:
- 1-48×2 ml tube; 1-24×4.5 ml tube; 1-12×15 ml tube; 2×50 ml tube
- Kit compatibility:
- All FastDNA Kits; All FastRNA Pro Kits; All FastRNA Spin Kits; All FastProtein Kits
- Key applications:
- Suitable for all sample types
Lysing highly contagious samples
Lysis of yeast and fungi
Used for isolation of PCR-ready genomic DNA from fecal samples, fungi and bacteria samples and also from soil samples
DNA & RNA purification
Lysis of bacteria
Lysis of soft tissues
Isolation of genomic DNA
Purification of DNA
Cell lysis
Used to lyse cell wall of yeast
Break hard/brittle samples
Used to isolate DNA from virtually any source
- Format:
- bead beating
- Adapter compatibility:
- Standard QuickPrep 24×2 ml tube holder; additional tube holders available:
QuickPrep 6002512 24×2 ml
BigPrep 6002525 2×50 ml
TeenPrep 6002526 12×15 ml
HiPrep 6002527 48×2 ml
CoolPrep (Cryogenic) 6002528 24×2 ml
CoolTeenPrep (Cryogenic) 6002530 6×15 ml
CoolBigPrep (Cryogenic) 6002531 2×50 ml
TallPrep 6002540 24×4.5 ml
All-Metal QuickPrep 6002545 24×2 ml
All-Metal TeenPrep 6002546 12×15 ml
All-Metal BigPrep 6002547 2×50 ml