Order Entry
Canada
ContactUsLinkComponent
Straight Inlet Liner with Wool for Thermo Scientific GCs, Restek
Straight Inlet Liner with Wool for Thermo Scientific GCs, Restek
  75779-526
 :  Restek
Straight Inlet Liner with Wool for Thermo Scientific GCs, Restek
  75779-526
 :  Restek
 :  23257

 

  • Ext.Ø:
    6.5 mm
  • Volume:
    0.986 ml
  • Description:
    Splitless, Straight with Base Deactivated Wool
  • Material:
    Borosilicate glass
  • Cat. no.:
    75779-526
  • For Use With:
    Thermo
  • I.D.×L:
    4.0×78.5 mm
  • Treatment:
    Base deactivated
  • Supplier No.:
    23257

 

 

Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.

  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination

Superior coverage is delivered even with highly active compounds.