Helium-Specific Replacement Triple Trap, Restek
Catalog # 10854-428
Supplier: Restek
Specifications
- Description:Helium-Specific Replacement Triple Trap (removes oxygen, moisture and hydrocarbons)
- Cat. no.:10854-428
- Supplier No.:22473
Specifications
About this item
This helium-specific triple trap effectively removes oxygen, moisture, and hydrocarbons from GCs.