ContactUsLinkComponent
Replacement Parts and Accessories for Mininert® Precision Sampling Valves for Vials, Restek
Replacement Parts and Accessories for Mininert® Precision Sampling Valves for Vials, Restek
Catalog # 10854-244
Supplier:  Restek
undefined
Replacement Parts and Accessories for Mininert® Precision Sampling Valves for Vials, Restek
Catalog # 10854-244
Supplier:  Restek
Supplier Number:  24907
Specifications
  • Description:
    Septum Insertion Tool
  • Cat. no.:
    10854-244
  • Supplier No.:
    24907
Specifications
About this item
These replacement parts for Mininert® Precision sampling valves for vials are extremely functional and economical.
  • Replacement septa are delivered in packs of 50
  • Septum insertion tool is made of durable materials
For use with Mininert® Precision sampling valves for vials.
Septa
Product Information