Single-Stage Ultra-High Purity Chrome-Plated Brass Gas Regulators with International Fittings, Restek
: Restek
UHP brass regulators are the best choice when using ultra-high-purity carrier gas for sensitive GC applications using MS, PID, or ECD detection methods.
- Use when there is secondary pressure regulation downstream
- Identical gas purity protection as with dual-stage gas regulators
- Oxidation-resistant
- Chrome-plated
All regulators are rated to 3000 psig maximum inlet pressure.
- Catalog No:
- 10854-748
- 10854-910
- 10854-630
- 10854-628
- 10854-624
- 10854-626
- Outlet gauge:
- 30", 0 to 14 bar (0 – 200 psig)
- 30", 0 to 14 bar (0 – 200 psig)
- 30", 0 to 14 bar (0 – 200 psig)
- 30", 0 to 14 bar (0 – 200 psig)
- 30", 0 to 14 bar (0 – 200 psig)
- 30", 0 to 14 bar (0 – 200 psig)
Frequently Bought Together