GRAPHPACK® 2M Ferrules for Gerstel CIS 3 and CIS 4 PTV Inlets, Restek
Supplier: Restek
Certificates
About this item
GRAPHPACK® technology was designed to provide leak-free, low dead volume, inert connections quickly and reliably for any type of connection used for gas chromatography.
- Available in a variety of pure or blended graphite and Vespel® fabrications
GRAPHPACK® 2M ferrules for micropacked columns are suitable for 1/16" fittings. Fit columns with internal diameters ranging from 0.25–0.53 mm.
Frequently Bought Together