Ferrules for Micropacked Columns, Compact—for Agilent 5890/6850/6890/7890 GCs, Restek
Supplier: Restek
Certificates
About this item
Ferrules are designed for use with micropacked columns in Agilent 5890/6850/6890/7890 GCs
- Graphite construction
Fit columns with internal diameters of 0.75 mm.
Ordering information: For micropacked columns.
Frequently Bought Together