Order Entry
Canada
ContactUsLinkComponent
Click-On In-line Super Clean® Ultra-High Capacity Oxygen Trap, Restek
Click-On In-line Super Clean® Ultra-High Capacity Oxygen Trap, Restek
Catalog # 10855-404
Supplier:  Restek
Click-On In-line Super Clean® Ultra-High Capacity Oxygen Trap, Restek
Catalog # 10855-404
Supplier:  Restek
Supplier Number:  22468

Specifications

  • Description:
    Ultra-High Capacity Oxygen Trap
  • Cat. no.:
    10855-404
  • Supplier No.:
    22468

Specifications

About this item

Click-on traps measure 21.6×3.2 cm (8.5×1.25").

  • Effective trap for catching oxygen
Ordering information: Replacement trap only - fittings sold separately.