ContactUsLinkComponent
Inlet Maintenance Kit for Agilent GCs, Restek
Inlet Maintenance Kit for Agilent GCs, Restek
Catalog # 10803-276
Supplier:  Restek
undefined
Inlet Maintenance Kit for Agilent GCs, Restek
Catalog # 10803-276
Supplier:  Restek
Supplier Number:  22181
Specifications
  • Description:
    Inlet Maintenance Kit
  • Cat. no.:
    10803-276
  • Supplier No.:
    22181
Specifications
About this item
For use with Agilent 5890, 6850, 6890, and 7890 GCs.
  • Kit includes the most common consumable GC supplies and tools
  • Parts list makes reordering easy
  • All parts meet or exceed performance of instrument manufacturer's parts