ContactUsLinkComponent
Inland 45 Pump Oil, Restek
Inland 45 Pump Oil, Restek
Catalog # 89205-674
Supplier:  Restek
undefined
Inland 45 Pump Oil, Restek
Catalog # 89205-674
Supplier:  Restek
Supplier Number:  24819
Specifications
  • Description:
    Inland 45 Pump Oil
  • Environmentally Preferable:
  • Cat. no.:
    89205-674
  • Supplier No.:
    24819
Specifications
About this item
Recommended for optimum mass spec performance, this oil pump is compatible with buna-N, neoprene, and Viton® seals.
  • Nontoxic
  • Noncorrosive
  • Ease at cold start
  • Optimum vacuum pump performance
This pump oil functions at low vapor pressure 10-7 torr.