Replacement Plunger-in-Needle Kits, SGE, Restek
Supplier: Restek
Certificates
About this item
Kits are supplied with both a plunger and a needle.
- Replacement plunger volume of 1 μL
- Needles are 70 mm in length
Ordering information: Plunger and needle must be replaced together.
Frequently Bought Together