Capillary Installation Gauge for Bruker/Varian GCs for use with ¹/₁₆" ferrules, Restek
Catalog # RK22335
Supplier: Restek
Specifications
- Description:Capillary Installation Gauge for use with Varian® GC, 1/16" Ferrules
- Cat. no.:RK22335
- Supplier No.:22335
Specifications
About this item
Use a capillary installation gauge for Bruker/Varian GCs to seat ferrules onto a column for consistent installations.
- For use with graphite ferrules only
Capillary installation gauges are for use with 1/16" ferrules.
Ordering information: For use with graphite ferrules only.