Septum Nut Removal Tool, Restek
Catalog # 10852-850
Supplier: Restek
Specifications
- Description:Septum Nut Removal Tool
- Cat. no.:10852-850
- For Use With:Agilent 5890/6850/6890/7890 GCs
- Supplier No.:24918
Specifications
About this item
Use this septum nut removal tool to easily remove the septum nut without touching the heated nut.
- Unique, ergonomic handle that is easy to grip
- Nut remains in tool for quick reattachment