Orders LinkContactUsLinkComponent
Wall Mounting Bracket, Restek
Wall Mounting Bracket, Restek
Wall Mounting Bracket for Super Clean Base Plates
Catalog # RK21984
Supplier:   Restek
Supplier #:  21984
Wall Mounting Bracket, Restek
Catalog # RK21984
Supplier:  Restek
Wall Mounting Bracket, Restek
Wall Mounting Bracket, Restek
Wall Mounting Bracket for Super Clean Base Plates
Price
$268.26
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Optional wall mounting bracket allows for the mounting of Super Clean® base plates with the use of screws.
  • Additional support for mounting base plates
Base plates can be mounted by using screws and the mounting holes on the base plate, or by using this optional wall mounting bracket.
Specifications
  • Description:
    Wall Mounting Bracket for Super Clean Base Plates