Order Entry
Canada
ContactUsLinkComponent
GC Oven Thermometer, Restek
GC Oven Thermometer, Restek
Catalog # 10852-898
Supplier:  Restek
undefined
GC Oven Thermometer, Restek
Catalog # 10852-898
Supplier:  Restek
Supplier Number:  22066

Specifications

  • Description:
    GC Oven Thermometer
  • Cat. no.:
    10852-898
  • Range:
    −50...+350 °C
  • Supplier No.:
    22066

Specifications

About this item

Oven thermometers can be used to verify GC oven temperature.

  • Quick measurements via sensor in the measuring tip
  • Swivel head can turn 180°
  • Wide measuring range: –50 to +350 °C