Order Entry
Canada
ContactUsLinkComponent
Shortix® Capillary GC Column Cutter, Restek
Shortix® Capillary GC Column Cutter, Restek
Catalog # 10852-828
Supplier:  Restek
undefined
Shortix® Capillary GC Column Cutter, Restek
Catalog # 10852-828
Supplier:  Restek
Supplier Number:  23026

Specifications

  • Description:
    Shortix Capillary GC Column Cutter
  • Cat. no.:
    10852-828
  • Supplier No.:
    23026

Specifications

About this item

Consistently make precise, clean, square cuts with a diamond blade.

  • Built-in magnifier to verify square cut

Use with tubing of 0.25 to 0.53 mm internal diameter (maximum 0.78 mm OD).

Accessories information: Maintenance kit for Shortix® capillary GC column cutter, includes diamond cutting wheel, O-rings, and a tool to open the column cutter.