ContactUsLinkComponent
ECD Vent Trap, Restek
ECD Vent Trap, Restek
  10855-828
 :  Restek
undefined
ECD Vent Trap, Restek
  10855-828
 :  Restek
 :  22017
  • Description:
    ECD Vent Trap, 1/8"
  • Cat. no.:
    10855-828
  • Supplier No.:
    22017
The ECD vent trap reduces the release of hazardous materials from the ECD vent into the lab.
 : Includes connecting lines and mounting kit.