Encapsulated Ferrules for ¹/₁₆" Compression Fittings, Restek
Supplier: Restek
Certificates
About this item
Encapsulated ferrules are reusable and will not deform and stick in fittings.
- Less torque needed to seal ferrule
- Restek’s unique blend of graphite minimizes fragmentation and out-gassing
For use with 1/16-inch compression fittings.
Frequently Bought Together