Inlet Liner Removal Tool, Restek
Inlet Liner Removal Tool
Catalog # RK20181
Supplier: Restek
Supplier #: 20181
Jump to:
Product Details & Documents
With this tool, easily remove liners from injectors without any more burned fingers.
- Made from high-temperature silicone
- Won’t chip or crack the liner
Specifications
- Description:Inlet Liner Removal Tool