Ultra-High Purity Chrome-Plated Brass Line Gas Regulator, Restek
Supplier: Restek
Certificates
About this item
Use these ultra-high purity line gas regulators where you need to reduce the line pressure by 20 psig (138 kPa) or more.
- Oxidation-resistant
- Chrome-plated brass fabrication
- Same purity protection as high-pressure cylinder regulators
Gas regulators feature 1/4" female NPT ports.
Ordering information: Order appropriate male connector, pipe-to-tube fittings.
Specifications
- Catalog No:
- 10855-354
- RK21666
- Outlet pressure:
- 0–150 psig
- 0–50 psig
- Inlet/outlet:
- 1/4" FPT/1/4" FPT Port
- 1/4" FPT/1/4" FPT Port
- Outlet gauge:
- 30", 0–200 psig (0–1,379 kPa)
- 30", 0–100 psig (0–689 kPa)