ContactUsLinkComponent
DI Liner Adapter for Agilent 5890 Packed Column GC, Restek
DI Liner Adapter for Agilent 5890 Packed Column GC, Restek
Catalog # RK21303
Supplier:  Restek
DI Liner Adapter for Agilent 5890 Packed Column GC, Restek
Catalog # RK21303
Supplier:  Restek
Supplier Number:  21303
Specifications
  • Description:
    DI Liner Adaptor for Agilent 5890 Packed Column GC
  • Cat. no.:
    RK21303
  • For Use With:
    for Agilent 5890 Packed Column GC
  • Supplier No.:
    21303
Specifications
About this item
Direct injection liner adapters use standard 1/16-inch capillary nuts and ferrules.
  • Convenient wrench pad at base
  • Includes 1/4-inch graphite ferrule and stainless steel nut
Use with Agilent or Restek DI glass inserts or Restek DI Uniliner® liners for an Agilent 5890 packed column GC.