ECD/FID Dual-Purpose Make-Up Gas Kit for Agilent 5890 GCs, Restek
Catalog # RK21301
Supplier: Restek
Specifications
- Description:ECD/FID Replacement Fitting Kit with Flow Manifold Connection
- Cat. no.:RK21301
- Supplier No.:21301
Specifications
About this item
ECD/FID dual-purpose make-up gas kits are for use with Agilent 5890 GCs.
- Kit includes: Replacement fitting, 1/4" nut, Vespel®/graphite ferrule, 1/16" nut, 0.4 mm ID graphite ferrule, Siltek®-treated guide
Kit includes: replacement fitting, 1/4" nut, Vespel®/graphite ferrule, 1/16" nut, 0.4 mm ID graphite ferrule, Siltek®-treated guide.