ContactUsLinkComponent
FID/NPD Adapter Fitting, Restek
FID/NPD Adapter Fitting, Restek
  10855-866
 :  Restek
undefined
FID/NPD Adapter Fitting, Restek
  10855-866
 :  Restek
 :  20884
  • Description:
    FID/NPD Adaptor Fitting
  • Cat. no.:
    10855-866
  • Supplier No.:
    20884
Includes 1/4 and 1/16" stainless steel nuts and 1/4" Vespel® and 0.4 mm ID graphite ferrules.
  • Easy-to-use, sturdy, compact stainless steel fitting
  • 1/16" nut uses standard graphite or Vespel®/graphite ferrules
  • Wrench pad won’t turn when installing a capillary column