Siltek® Septum Packed Purge Port Weldment for Agilent 5890 GCs, Restek
Catalog # RK21691
Supplier: Restek
Specifications
- Description:Siltek Septum Packed Purge Port Weldment for Agilent 5890 GCs
- Cat. no.:RK21691
- For Use With:for Agilent 5890 GCs
- Supplier No.:21691
Specifications
About this item
Siltek® treatment makes weldment inert and eliminates adsorption of sensitive compounds (e.g., DDT and endrin).
- Inert
- Eliminate adsorption of sensitive compounds
For use with Agilent 5890 GCs.
Ordering information: Order Viton® O-rings and appropriate septa separately.