¹/₈" Capillary Inlet Adapter Fitting Kit, Split/Splitless Fitting for 0.53 mm Int.Ø Capillary Columns, Restek
Catalog # RK20645
Supplier: Restek
Specifications
- Description:1/8-Inch Capillary Inlet Adaptor Fitting Kit
- Cat. no.:RK20645
- Supplier No.:20645
Specifications
About this item
1/8-inch split/splitless fitting accepts standard two-hole capillary ferrules and a standard 1/8-inch nut.
- Makes column installation easy due to the nut protruding farther from the insulated injection port chamber
- The column insertion depth is the same as the original equipment
- Kit includes adapter fitting, capillary nut, stainless steel inlet seal, washer, and one 0.8 mm ID two-hole ferrule
- Use recessed taper liners with this adapter
These inlet adapter fitting kits make column installation easy with far-protruding nuts.