Polyimide, resin
5 g
Catalog # RK20445
Supplier: Restek
: 20445
Jump to:
Product Details & Documents
Resin securely connects a Press-Tight® connector to a fused silica column.
- Max operating temperature of 350°C
- Polyimide
- CAS Number:62929-02-6
- Formula:C₃₅H₂₈N₂O₇
- MDL Number:MFCD01868102
- Molecular Weight:588.616 g/mol
Specifications
Specifications
- Size:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



