ContactUsLinkComponent
Mini Wool Puller/Inserter, Restek
Mini Wool Puller/Inserter, Restek
Catalog # RK20114
Supplier:  Restek
undefined
Mini Wool Puller/Inserter, Restek
Catalog # RK20114
Supplier:  Restek
Supplier Number:  20114
Specifications
  • Description:
    Mini Wool Puller/Inserter
  • Cat. no.:
    RK20114
  • Supplier No.:
    20114
Specifications
About this item
This miniature puller/inserter device is ideal for handily manipulating wool plugs.
  • Slender design
  • Easy to maneuver
Insert and remove wool plugs easily.
Caution: Not recommended for use with double gooseneck liners.