ContactUsLinkComponent
Tapered Needle Files 4", Restek
Tapered Needle Files 4", Restek
Catalog # RK20106
Supplier:  Restek
undefined
Tapered Needle Files 4", Restek
Catalog # RK20106
Supplier:  Restek
Supplier Number:  20106
Specifications
  • Description:
    4" Tapered Needle Files
  • Cat. no.:
    RK20106
  • Supplier No.:
    20106
Specifications
About this item
Tapered needle files serve a variety of uses where fine points of manipulation are required.
  • Very fine point
  • Slender design
  • Durable fabrication
These files can be used for many purposes. They are especially useful for removing ferrules that are lodged in injector or detector ports.