ContactUsLinkComponent
Slide-Lock Tweezers and 15 cm Compact Steel Ruler, Restek
Slide-Lock Tweezers and 15 cm Compact Steel Ruler, Restek
Catalog # RK20101
Supplier:  Restek
undefined
Slide-Lock Tweezers and 15 cm Compact Steel Ruler, Restek
Catalog # RK20101
Supplier:  Restek
Supplier Number:  20101
Specifications
  • Description:
    Slide-Lock Tweezers and 15 cm Compact Steel Ruler
  • Cat. no.:
    RK20101
  • Supplier No.:
    20101
Specifications
About this item
This duo provides an alternative way to install capillary columns to the correct insertion distance recommended by the instrument manufacturer.
  • Useful for many laboratory tasks
  • Durable steel construction
  • Easily install capillary columns
This duo provides an alternative way to install capillary columns to the correct insertion distance recommended by the instrument manufacturer.