Soap Film Bubble Flowmeter, Restek
Supplier: Restek
Certificates
About this item
- 1 mL flowmeter for flows between 0.1 and 10 cc/min
- 50 mL flowmeter for flows between 10 and 300 cc/min
Delivery: Includes a reservoir bulb, twenty-four inches (60 cm) of 1/4" ID tubing, adapter tubes for 1/8" tubing and 0.53 mm ID capillary columns, and Velcro® fasteners.
Customers Who Bought This Also Bought