ContactUsLinkComponent
Gas Management System, Restek
  10854-934
 :  Avantor
undefined
Gas Management System, Restek
  10854-934
 :  Avantor
  • Description:
    Restek Gas Management System
  • Cat. no.:
    10854-934
  • Includes:
    Fittings for 1/8" and 1/4" gas line
  • Supplier No.:
    21999
Removes moisture, hydrocarbons, and oxygen from carrier gas, extending column lifetime.
  • Produces high-purity carrier gas for most applications
  • Includes one each: moisture, hydrocarbon, and indicating oxygen trap
  • Replacing traps is safe and easy
  • Maximum flow: 150 cc/min for optimal performance
Restek has put together a convenient unit providing gas purification all in one step. Complete with an indicating trap, your gas purification issues are handled in one central location.
 : Items cannot ship FedEx ground.