Rotor Stand, Thermo Scientific
Catalog #: 89218-302
Supplier: Thermo Fisher Scientific
Specifications
- Description:Rotor Stand for Autolock Rotors
- Cat. no.:89218-302
- Supplier No.:75003711
Specifications
About this item
This rotor stand allows rotors and buckets to be removed and kept simply and safely.
Also suitable to store rotor when not in use.
Customers Who Bought This Also Bought