Orders LinkContactUsLinkComponent
Corning® CoolCell® Containers, Corning
Corning® CoolCell® Containers, Corning
Supplier:   Corning
Corning® CoolCell® Containers, Corning
Supplier:  Corning
Certificates
Corning® CoolCell® Containers, Corning
Corning® CoolCell® Containers, Corning
13 Options Available Below
Product Details & Documents
CoolCell® alcohol-free cell freezing containers ensure standardized controlled-rate −1 °C/minute cell freezing in a −80 °C freezer, without alcohol or any fluids.
  • No alcohol. no fluids, no pre-cooling
  • No variability, all vials have uniform freeze rate
  • Radially symmetric design ensures vial consistency
  • No on-going cost, no alcohol purchase or disposal
  • No stuck lids, ergonomic lid comes off easily when frozen
  • Not cold to the touch when removing from the −80 ˚C freezer
  • Quick re-use time, ready to use again after five minutes
Proven for use with a variety of cell types including stem cells, primary cells, PBMC cell lines, insect cells, yeast and others. The patent-pending CoolCell® technology utilizes a thermo-conductive alloy core and highly-insulative outer material to control the rate of heat removal and provide reproducible cell cryopreservation. CoolCell units are easy to use and deliver comparable results to expensive programmable freezers.
CoolCell® LX
For 12 standard 1,0 to 2,0 ml cryogenic vials, 1,0 ml fill per vial. Radially symmetric for uniform vial freezing. Numbered wells for easy sample identification. Beveled lid for secure gripping and easy opening. Exposed vial tops when lid is open for quick, organized removal of frozen samples.
CoolCell® FTS30
For 30 standard 1,0 to 2,0 ml cryogenic vials, 1,0 ml fill per vial. Controlled micro-convection for uniform freezing of 30 vials. Removable vial tray for one-step transfer of samples into and out of freezing chamber."
CoolCell® 5 mL LX
For 12 standard 3,5 to 5,0 ml fill cryogenic vials, 3,5 to 5,0 fill per vial. Radially symmetric for uniform vial freezing. Numbered wells for easy sample identification. Beveled lid for secure gripping and easy opening. Exposed vial tops when lid is open for quick, organized removal of frozen samples.
CoolCell® SV2 and CoolCell® SV10
For 12 standard 2,0 ml injectable ampules, 1,0 ml fill per ampule (SV2). For 6 standard 10,0 ml injectable ampules, 5,0 ml fill per ampule (SV10). Radially symmetric for uniform freezing of injectable ampules. Easy open lid. Exposed vial tops when lid is open for quick, organized removal of frozen samples.
CoolCell SV10 Stem Cell Cryopreservation System
includes the XT Starter Benchtop Cooler, Corning® CoolRack SV10 and CoolCell SV10 Cell Freezing Container for 6 injectable vials.
CoolCell® Filler Vials
To ensure cell freezing rate consistency and uniform results when using CoolCell containers, insert a CoolCell Filler Vial into empty wells when freezing less than a full load. Suitable for repeated use and compatible with CoolCell LX, CoolCell FTS30 and CoolCell 5 ml LX containers.
CoolCell® FTS30 Vial Module
FTS30 Vial Module is a holder for 30×1,0 ml or 2,0 ml cryogenic vials that allows one-step insertion and removal of all 30 vials at once. Fits into a standard 5,0×5,0×2,0" cryostorage box. Compatible with dry ice and liquid nitrogen.
Product Family Options
Product Information
  • No. of Holes
    Color
    For vials
    Well diameter
    Dimensions
    Availability
    Price
  • 75779-712
    CoolCell®, Alcohol-free Cell Freezing Container, Fully Covered Vial Tops
    CoolCell®, Alcohol-free Cell Freezing Container, Fully Covered Vial Tops
    12
    Purple
    1 or 2 ml cryogenic vials
    12.7 mm
    11.7×10.9 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $222.70
    $222.70
    New Customer Promotional Price
    Case of 1
       $222.70
    Supplier Number: 432000
  • 75779-716
    CoolCell® LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    CoolCell® LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    12
    Green
    1 or 2 ml cryogenic vials
    12.7 mm
    11.7×10.9 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $274.44
    $274.44
    New Customer Promotional Price
    Case of 1
       $274.44
    Supplier Number: 432002
  • 75779-718
    CoolCell® LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    CoolCell® LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    12
    Orange
    1 or 2 ml cryogenic vials
    12.7 mm
    11.7×10.9 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $278.80
    $278.80
    New Customer Promotional Price
    Case of 1
       $278.80
    Supplier Number: 432003
  • 75779-720
    CoolCell® LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    CoolCell® LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    12
    Pink
    1 or 2 ml cryogenic vials
    12.7 mm
    11.7×10.9 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $292.54
    $292.54
    New Customer Promotional Price
    Case of 1
       $292.54
    Supplier Number: 432004
  • 75779-714
    CoolCell® LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    CoolCell® LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    12
    Purple
    1 or 2 ml cryogenic vials
    12.7 mm
    11.7×10.9 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $278.80
    $278.80
    New Customer Promotional Price
    Case of 1
       $278.80
    Supplier Number: 432001
  • 75779-722
    CoolCell® 5 mL LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    CoolCell® 5 mL LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    12
    Purple
    3.5 - 5 ml cryogenic vials
    12.7 mm
    9.5×14.5 cm
    Log in for availability
    Log in for availability
    Case (5ml)
    Case (5ml) - $372.30
    $372.30
    New Customer Promotional Price
    Case (5ml)
       $372.30
    Supplier Number: 432005
  • 75779-816
    CoolCell® FTS30, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    CoolCell® FTS30, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    30
    Green
    1 or 2 ml cryogenic vials
    12.7 mm
    16.5×11.5 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $761.60
    $761.60
    New Customer Promotional Price
    Case of 1
       $761.60
    Supplier Number: 432008
  • 75779-814
    CoolCell® FTS30 Alcohol-free Cell Freezing Container, Exposed Vial Tops
    CoolCell® FTS30 Alcohol-free Cell Freezing Container, Exposed Vial Tops
    30
    Orange
    1 or 2 ml cryogenic vials
    12.7 mm
    16.5×11.5 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $761.60
    $761.60
    New Customer Promotional Price
    Case of 1
       $761.60
    Supplier Number: 432007
  • 75779-818
    CoolCell® FTS30, Alochol-free Cell Freezing Container, Exposed Vial Tops
    CoolCell® FTS30, Alochol-free Cell Freezing Container, Exposed Vial Tops
    30
    Pink
    1 or 2 ml cryogenic vials
    12.7 mm
    16.5×11.5 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $761.60
    $761.60
    New Customer Promotional Price
    Case of 1
       $761.60
    Supplier Number: 432009
  • 75779-724
    CoolCell® FTS30, Alcohol-fee Cell Freezing Container, Exposed Vial Tops
    CoolCell® FTS30, Alcohol-fee Cell Freezing Container, Exposed Vial Tops
    30
    Purple
    1 or 2 ml cryogenic vials
    12.7 mm
    16.5×11.5 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $761.60
    $761.60
    New Customer Promotional Price
    Case of 1
       $761.60
    Supplier Number: 432006
  • 75779-820
    CoolCell®SV2 Alcohol-free Freezing Container, Exposed Vial Tops
    CoolCell®SV2 Alcohol-free Freezing Container, Exposed Vial Tops
    12
    Purple
    2 ml serum vials
    14.7 mm
    13.9×10.5 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $532.95
    $532.95
    New Customer Promotional Price
    Case of 1
       $532.95
    Supplier Number: 432010
  • 75779-822
    CoolCell® SV10 Alchol-free Freezing Container, Exposed Vial Tops
    CoolCell® SV10 Alchol-free Freezing Container, Exposed Vial Tops
    6
    Purple
    10 ml serum vials
    23.6 mm
    12.1×9.8 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $532.95
    $532.95
    New Customer Promotional Price
    Case of 1
       $532.95
    Supplier Number: 432011
  • 76180-494
    CoolCell® LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    CoolCell® LX, Alcohol-free Cell Freezing Container, Exposed Vial Tops
    12
    Purple, Green, Orange, Pink
    1 or 2 ml cryogenic vials
    12.7 mm
    11.7×10.9 cm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $1,073.80
    $1,073.80
    New Customer Promotional Price
    Case of 1
       $1,073.80
    Supplier Number: 432138
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Removable Cryogenic vial Module for CoolCell FTS30, 3/Pk.
    Removable Cryogenic vial Module for CoolCell FTS30, 3/Pk.
    75779-782
    Log in for availability
    Case of 10 - $165.75
Product Information
  • Catalog #
    Availability
    Price
  • CoolCell® 2 mL Filler Vials for Use with CoolCell LX and CoolCell FTS30 Containers
    CoolCell® 2 mL Filler Vials for Use with CoolCell LX and CoolCell FTS30 Containers
    75779-778
    Log in for availability
    Case of 6 (2ml) - $46.20
Product Information
  • Catalog #
    Availability
    Price
  • Cryogenic Vial Grippers, Pack of 5, Assorted Colors
    Cryogenic Vial Grippers, Pack of 5, Assorted Colors
    75779-848
    Log in for availability
    Case of 5 - $83.56
Product Information
  • Catalog #
    Availability
    Price
  • CoolCell®5 mL Filler Vials for Use with CoolCell 5 mL LX Container
    CoolCell®5 mL Filler Vials for Use with CoolCell 5 mL LX Container
    75779-780
    Log in for availability
    Case of 6 (5ml) - $39.27