Orders LinkContactUsLinkComponent
Student Dissecting Scissors
Student Dissecting Scissors
Supplier:   Dunrite Instruments
Student Dissecting Scissors
Certificates
Student Dissecting Scissors
2 Options Available Below
Product Details & Documents
Great for general purpose dissection.
Great For General Purpose Dissection. These stainless Dissecting scissors will give you many years of service. Available in regular or Fine point.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Product Information
  • Length
    Material
    Blade type
    Availability
    Price
  • 470005-458
    Dissecting Scissors
    Dissecting Scissors
    115 mm (4.5")
    Stainless steel
    Sharp/Sharp
    Log in for availability
    Log in for availability
    Each
    Each - $8.70
    $8.70
    Each
       $8.70
    Supplier Number: 140601
  • 470122-008
    Dissection Scissors Fine Point
    Dissection Scissors Fine Point
    115 mm (4.5")
    Stainless steel
    Sharp/Sharp
    Log in for availability
    Log in for availability
    Each
    Each - $10.80
    $10.80
    Each
       $10.80
    Supplier Number: S26930