Specifications
- Length:150 mm
- Temperature range:0...80 °C
- Cat. no.:34117-944
- Internal Reference:Ag/AgCl
- Junction:Ceramic
- Supplier No.:9780SC
Specifications
About this item
Obtain an accurate determination of potentiometric halides with dual electrodes using these silver billet electrodes, these electrodes are a combination of a silver billet-indicating electrode along with a silver billet half-cell.